Learn More
CD162 (PSGL-1) Monoclonal Antibody (FLEG), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 12162942
Description
Description: This FLEG monoclonal antibody recognizes human CD162, which is also known as P-selectin glyocoprotein ligand-1 (PSGL-1). This 120-kDa protein exists as a homodimer on the surface of monocytes, neutrophils, granulocytes, peripheral T cells, some B cells, and a subset of CD34+ hematopoietic progenitor cells (HPCs) in the bone marrow. CD162 binds CD62P (P-selectin), CD62E (E-selectin), and CD62L (L-selectin) to mediate leukocyte interactions with each other as well as with activated platelets and endothelium. Thus, CD162 plays a critical role in leukocyte adhesion and rolling. Furthermore, the binding of CD162 to P-selectin on HPCs has been demonstrated to inhibit hematopoiesis. Studies have also demonstrated CD162 as the receptor for enterovirus 71. Applications Reported: This FLEG antibody has been reported for use in flow cytometric analysis. Applications Tested: This FLEG antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.Specifications
CD162 (PSGL-1) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q14242 | |
SELPLG | |
Affinity chromatography | |
RUO | |
6404 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
FLEG | |
PE | |
SELPLG | |
CD162; CLA; Cutaneous lymphocyte associated antigen; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; Psgl-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.