Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD162 (PSGL-1) Rat anti-Mouse, PE, Clone: 4RA10, Invitrogen™

Rat Monoclonal Antibody

$158.00 - $366.00

Specifications

Antigen CD162 (PSGL-1)
Clone 4RA10
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12-162-180
Encompass
View Documents
Invitrogen
12162180
25 μg
Each for $158.00
Add to cart
 
12-162-182
Encompass
View Documents
Invitrogen
12162182
100 μg
Each for $366.00
Add to cart
 
Description

Description

Description: This 4RA10 monoclonal antibody reacts with mouse CD162, which is also known as P-selectin glycoprotein ligand-1 (PSGL-1). This clone has been successfully used in flow cytometry and has been reported to block binding of CD162 to CD62P and CD62L. Applications Reported: This 4RA10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4RA10 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser

CD162 (PSGL-1) is a 120 kDa homodimer protein found on the surface of leukocytes and platelets. CD162 is known to bind CD62P (P-selectin), CD62L (L-selectin), and to a lower degree CD62E (E-selectin). These interactions play a critical role in leukocyte adhesion, rolling and extravasation. CD162 has been shown to be an important immune marker of T cell exhaustion in chronic viral infections and cancer.
Specifications

Specifications

CD162 (PSGL-1)
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
CD162; CLA; cutaneous lymphocyte-associated associated antigen; leukocyte cell surface adhesion molecule; P-selectin glycoprotein ligand 1; P-selectin glycoprotein ligand 1 propeptide; P-selectin glycoprotein ligand-1; Psgl1; PSGL-1; Selectin P ligand; selectin, platelet (p-selectin) ligand; Selp1; SELPG; Selpl; SELPLG
SELPLG
IgG1 κ
Affinity chromatography
Antibody
4RA10
Flow Cytometry
PE
Rat
Mouse
Q62170
20345
Mouse PSGL-1 human IgG1 fusion protein
Primary
4°C, store in dark, DO NOT FREEZE!
SELPLG
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit