Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Antibody (GHI/61), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143341APC
Description
CD163 Monoclonal specifically detects CD163 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
CD163 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
CD163 | |
The immunogen for this antibody was CD163. | |
0.1 mL | |
Immune System Diseases, Immunology, Innate Immunity | |
9332 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
GHI/61 | |
Flow Cytometry | |
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction