Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Mouse anti-Human, Clone: 2G12, Abnova™

Mouse monoclonal antibody raised against full length recombinant human CD163.

Manufacturer:  Abnova Corporation MAB13002

Catalog No. 89-933-746


Add to cart

Description

Description

Specifications

Specifications

CD163
2G12
Mouse monoclonal antibody raised against full length recombinant human CD163.
In PBS, pH 7.3 (50% glycerol, 1% BSA, 0.02% sodium azide).
M130/MM130
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
CD163
Recombinant protein corresponding to full length human CD163.
Protein A/G purification
RUO
Primary
9332
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit