Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Mouse anti-Human, PerCP, Clone: GHI/61, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143341PCP

Catalog No. NBP143341JP


Add to cart

Description

Description

CD163 Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Flow Cytometry, Western Blotting.
Specifications

Specifications

CD163
GHI/61
Western Blot, Flow Cytometry
Purified
CD163
The immunogen for this antibody was CD163.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
PerCP
PBS with 0.05% Sodium Azide
CD163 antigenscavenger receptor cysteine-rich type 1 protein M130, CD163 molecule, Hemoglobin scavenger receptor, M130, M130macrophage-associated antigen, MM130
Mouse
IgG1 Kappa
0.1 ml
Immune System Diseases, Immunology, Innate Immunity
Primary
9332
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit