Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD163 Polyclonal Antibody, Huabio
Rabbit Polyclonal Antibody
Supplier: HUABIO RESEARCH INC ER200115


Description
CD163 Polyclonal antibody specifically detects CD163 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
CD163 | |
Polyclonal | |
Unconjugated | |
CD163 | |
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
Synthetic peptide within human CD163 aa 42-90. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q86VB7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9332 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction