Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Rabbit anti-Human, Clone: M130/3708R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30757210UG

 View more versions of this product

Catalog No. NB122617


Add to cart

Description

Description

CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD163
M130/3708R
Unconjugated
10 mM PBS with 0.05% BSA
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Synthetic peptide corresponding to CD163 residues within aa1056-1156 of CD163 was used as an immunogen.
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin)
0.2 mg/ml
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Rabbit
IgG Kappa
100 μg
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
Primary
9332
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit