Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD163 Rabbit anti-Human, Clone: M130/3708R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen CD163
Clone M130/3708R
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122618
View Documents
Novus Biologicals
NBP30757220UG
20 μg
Each for $159.82
Add to cart
 
NB122617
View Documents
Novus Biologicals
NBP30757210UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

CD163 Monoclonal antibody specifically detects CD163 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

CD163
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Purified
RUO
Human
CD_antigen: CD163, CD163, CD163 molecule, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163
Synthetic peptide corresponding to CD163 residues within aa1056-1156 of CD163 was used as an immunogen.
Monoclonal
Protein A or G purified
M130/3708R
ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Angiogenesis, Immune System Diseases, Immunology, Inflammation, Innate Immunity
10 mM PBS with 0.05% BSA
9332
IgG Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit