Learn More
CD167a Mouse anti-Human, Alexa Fluor 647, Clone: 48B3, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566236
Catalog No. BDB566236
Add to cart
Description
Description
CD167a Monoclonal antibody specifically detects CD167a in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD167a
48B3
Alexa Fluor 647
Purified
Mouse
Mouse BALB/c IgM, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions, and unreacted Alexa Fluor™ 647 was removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
CD167; CAK; DDR; EDDR1; HGK2; MCK10; NEP; NTRK4; PTK3; PTK3A; RTK6; TRKE
Human DDR1 Transfected Cell Line
50μg
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.