Learn More
CD169 Rat anti-Mouse, Brilliant Violet 421, Clone: 3D6/CD169, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566604
Catalog No. BDB566604
Add to cart
Description
Description
CD169 Monoclonal antibody specifically detects CD169 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD169
3D6/CD169 (also known as 3D6)
Brilliant Violet 421
Rat
0.1 mg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2a, κ
RUO
Primary
CD169