Learn More
CD16b Mouse anti-Human, BV605, Clone: CLB-gran11.5, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745143
Catalog No. BDB745143
Add to cart
Description
Description
CD16b Monoclonal antibody specifically detects CD16b in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD16b
CLB-gran11.5
Brilliant Violet 605
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Human