Learn More
CD16b Mouse, anti-Human, R718, Clone: CLB-GRAN11.5, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 751880
Catalog No. BDB751880
Add to cart
Description
Description
Reacts with CD16b, a glycosyl phosphatidyl inositol-anchored (GPI) protein expressed on human neutrophils. Human CD16 is the low affinity Fc gamma receptor III (Fc gamma RIII) and shows two distinct forms coded by two linked genes. One form is CD16a, a polypeptide-anchored form (Fc gamma RIIIA), present on natural killer cells and macrophages. The other form is CD16b the GPI-anchored form, Fc gamma RIIIB, found on human neutrophils. CD16b is polymorphic and the two codominant alleles are referred to as Neutrophil Antigen 1 (NA1) and Neutrophil Antigen 2 (NA2). Clone CLBgran11.5 reacts with neutrophils expressing the NA1 molecule. CD16b has been reported to participate in immune complex binding by resting neutrophils and also in neutrophil transendothelial migration by interacting with integrins during the inflammation response process.
Specifications
Specifications
CD16b
CLB-GRAN11.5
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human