Learn More
CD172a/b Mouse anti-Human, Brilliant Violet 510, Clone: SE5A5, BD Optibuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 743563
Description
The SE5A5 monoclonal antibody specifically binds to a common epitope on CD172a/ SIRPα (90 kDa) and CD172b/ SIRPβ 1 (50 kDa) . These transmembrane glycoproteins are members of the Signal Regulatory Protein (SIRP) family that, in turn, belongs to the Immunoglobulin superfamily. The SIRP family is comprised of two subgroups, SIRPα and SIRPβ that are distinguished by the presence (α) or absence (β) of a cytoplasmic domain containing two immunoreceptor tyrosine-based inhibition motifs (ITIM). CD172a/ SIRPα is expressed on CD34+ stem/progenitor cells, cardiomyocytes, monocytes, macrophages, granulocytes, dendritic cells, and in the central nervous system. It binds to CD47 and is implicated in mediating inhibitory signals via the ITIM/SHP-2 association. CD172b/ SIRPβ 1 does not possess a cytoplasmic domain but instead the transmembrane domain contains a positively-charged residue that can interact with another transmembrane protein, DAP-12/KARAP. DAP-12 has two immunoreceptor tyrosine-based activat ion motifs (ITAM) within its cytoplasmic domain that are thought to link CD172b to cellular activation signaling . CD172b is expressed on myeloid cells, including peripheral blood monocytes and granulocytes. It is not expressed on CD34+ cells. CD172a and CD172b have complementary role s in signal regulation and may work together in tuning certain cellular responses to stimuli.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.Specifications
CD172a/b | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
SIRP alpha/beta1, SIRPα/SIRPβ1, Signal Regulatory Protein α/β1 | |
Human SIRP alpha extracellular domain Recombinant Protein | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV510 under optimal conditions that minimize unconjugated dye and antibody. | |
RUO | |
Primary | |
Human |
SE5A5 | |
Brilliant Violet 510 | |
Affinity Purified | |
Murine | |
Mouse BALB/c IgG1, κ | |
50μg | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
Monoclonal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.