Learn More
CD177 Mouse anti-Human, FITC, Clone: MEM-166, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen A15743
Catalog No. 50-113-7742
Add to cart
Description
Description
CD177 (NB1/HNA-2a and PRV-1 form) is a GPI-anchored glycoprotein present mainly on neutrophils. Its plasma membrane expression is increased during pregcy and and inflammation or after G-CSF application. Ligand of CD177 has been identified as CD31 (PECAM-1). CD177 participates in neutrophil transmigration and seems to be also a pro-proliferative molecule. The antibodies against CD177 can be involved in neonatal alloimmune neutropenia (NAN).
Specifications
Specifications
CD177
Monoclonal
FITC
CD177
Liquid
CD177
Human CD177
Purified
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MEM-166
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Q8N6Q3
HNA-2a, HNA2A, NB1, NB1 GP, PRV-1, PRV1, CD177 antigen, NB1 glycoprotein, cell surface receptor, human neutrophil alloantigen 2a, polycythemia rubra vera 1, polycythemia rubra vera protein 1
Mouse
IgG1
100 tests
4° C
Primary
57126