Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD178 (Fas Ligand) Armenian Hamster anti-Mouse, PE, Clone: MFL3, eBioscience™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen 12591183

 View more versions of this product

Catalog No. 50-109-90


Add to cart

Description

Description

Description: The MFL3 monoclonal antibody reacts with mouse Fas (CD95) Ligand, a 40 kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by mouse activated T cells. The interaction of FasL with its receptor CD95 induces Fas-mediated killing. It has been reported that the human FasL antigen is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26 kDa soluble form. The degree of sensitivity for the mouse antigen to MMPs has not been reported. Applications Reported: The MFL3 antibody has been reported for use in flow cytometric analysis. Applications Tested: The MFL3 antibody has been tested by flow cytometric analysis of mouse FasL transfected cells and activated T cells. This can be used at less than or equal to 1 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 µm post-manufacturing filtered.

Fas ligand (FasL, CD95L) is a type-II-membrane protein, whose N-terminus is in the cytoplasm and its C-terminal region extends into the extracellular space. Its receptor Fas (CD95, Apo-1) is a cell-surface-type-I-membrane protein and a member of the tumor necrosis factor (TNF) and nerve growth factor (NGF) receptor family. As a member of the TNF-cytokine family FasL induces apoptosis when interacting with its receptor Fas. FasL may exist as either membrane bound (45 kD) or soluble forms (26 kD). The soluble protein can be released from cells upon cleavage by metalloproteinases. Binding of FasL to Fas leads to oligomerization of the receptor and triggers apoptotic cell death through the interaction of other proteins. FasL is predomitly expressed in activated T-lymphocytes and natural killer (NK) cells also it is expressed in the tissues of immune-privilege sites such as testis and eye. FasL expression is also reported in various tissues as thymus, liver, ovary, lung, heart and kidney. The Fas/FasL system has been shown to play a role in a number of human diseases, for example AIDS, hepatitis or cancer. It is assumed that induction of apoptosis through FasL is predomitly involved in anti-viral immune responses.
Specifications

Specifications

CD178 (Fas Ligand)
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P41047
FasL, apoptosis antigen ligand 1, CD95 ligand, CD95L, CD178, APO1, Fas Ligand, Fas-LG
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
MFL3
PE
Fasl
Liquid
Fasl
IgG
200 μg
4° C, store in dark, DO NOT FREEZE!
Primary
14103
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.