Learn More
CD178 (Fas Ligand) Monoclonal Antibody (MFL3), PE-Cyanine7, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$341.00
Specifications
Antigen | CD178 (Fas Ligand) |
---|---|
Clone | MFL3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-9206
|
Invitrogen
25591182 |
100 μg |
Each of 1 for $341.00
|
|
|||||
Description
Description: The MFL3 monoclonal antibody reacts with mouse Fas (CD95) Ligand, a 40 kDa type II transmembrane glycoprotein. FasL is a member of the TNF family and is expressed by mouse activated T cells. The interaction of FasL with its receptor CD95 induces Fas-mediated killing. It has been reported that the human FasL antigen is cleaved from the surface by matrix metalloproteinases (MMPs), resulting in a 26 kDa soluble form. The degree of sensitivity for the mouse antigen to MMPs has not been reported. Applications Reported: This MFL3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MFL3 antibody has been tested by flow cytometric analysis of transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD178 (Fas Ligand) Monoclonal antibody specifically detects CD178 (Fas Ligand) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD178 (Fas Ligand) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
ADAM10-processed FasL form; ALPS1B; APL; Apoptosis (APO 1) antigen ligand 1; apoptosis (APO-1) antigen ligand 1; Apoptosis (APO-1) antigen ligand 1 (Fas antigen ligand); apoptosis antigen ligand; APT1LG1; APTL; CD178; CD178 antigen; CD95 ligand; CD95 ligand protein;Generalized lymphoproliferative disease (Gld); Cd95l; CD95-L; CD95L protein; fas antigen ligand; Fas ligand; Fas ligand (TNF superfamily, member 6); Fasl; Fas-L; FasL ICD; FasL intracellular domain; Faslg; Fas-LG; generalized lymphoproliferative disease; gld; mutant tumor necrosis factor family member 6; Receptor-binding FasL ectodomain; sFasL; Soluble Fas ligand; soluble form; SPA; SPPL2A-processed FasL form; TNFL; Tnfsf6; TNLG1A; Tumor necrosis factor (ligand) superfamily member 6; Tumor necrosis factor (ligand) superfamily member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor (ligand) superfamily, member 6 (apoptosis (APO-1) antigen ligand 1) (Fas antigen ligand); Tumor necrosis factor | |
Fasl | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Fasl |
MFL3 | |
Flow Cytometry | |
PE-Cyanine7 | |
Armenian Hamster | |
Mouse | |
P41047 | |
14103 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.