Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD18 Rat anti-Mouse, BUV395, Clone: GAME-46, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 744485
Description
CD18 Monoclonal antibody specifically detects CD18 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD18 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
GAME-46 | |
Brilliant Ultraviolet 395 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction