Learn More
CD182 (CXCR2) Mouse, anti-Mouse, R718, Clone: V48-2310, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752139
Catalog No. BDB752139
Add to cart
Description
Description
The V48-2310 monoclonal antibody specifically recognizes CD182 which is also known as C-X-C chemokine receptor type 2 (CXCR2). CD182 is a seven-transmembrane protein and member of the G protein-coupled receptor 1 family that is encoded by Cxcr2 [Chemokine (C-X-C motif) receptor 2]. CD182 is highly expressed on neutrophils and binds to CXCL1 (KC/GRO-alpha), CXCL2 (MIP2), CXCL3 (Dcip1), CXCL5 (LIX), and CXCL7 (NAP-2). CD182 plays a role in neutrophil activation and trafficking.
Specifications
Specifications
CD182 (CXCR2)
V48-2310
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Mouse