Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD183 Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma, BB700, Clone: 1C6/CXCR3 (also known as 1C6, LS177-1C6), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566532
Description
CD183 Monoclonal antibody specifically detects CD183 in Human,Baboon,Cynomolgus,Rhesus Ma samples. It is validated for Flow Cytometry.Specifications
CD183 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Flow Cytometry | |
1C6/CXCR3 (also known as 1C6, LS177-1C6) | |
Mouse | |
RUO | |
Human, Baboon, Cynomolgus, Rhesus Macaque | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction