Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD185 (CXCR5) Rat anti-Mouse, BB700, Clone: 2G8, BD Horizon™

Rat Monoclonal Antibody

$160.00 - $385.00

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen CD185 (CXCR5)
Conjugate BB700
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB566493
View Documents
BD Biosciences
566493
25 μg Each for $160.00
Add to cart
 
BDB566492
View Documents
BD Biosciences
566492
0.1 mg Each for $385.00
Add to cart
 
Description

Description

CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Monoclonal
Primary
CD185 (CXCR5)
BB700
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit