Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19, anti-Human, Clone: 4G7, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00121

Catalog No. 89-032-464


Add to cart

Specifications

Specifications

CD19
4G7
Flow Cytometry (1 ug/mL) Immunofluorescence The optimal working dilution should be determined by the end user.
CD19 molecule
B4|MGC12802
Original antibody is raised against human chronic lymphocytic leukemia cells.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
930
Flow Cytometry, Immunofluorescence
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
CD19
IgG, kappa
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit