Learn More
CD19 Monoclonal Antibody (SJ25-C1), PE, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Invitrogen 502544952
Encompass_Preferred
Catalog No. 50-254-4952
Explore available promotions
Add to Cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.