Learn More
CD19 Mouse anti-Human, Alexa Fluor™ 350, Clone: C19/366, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233194AF350
Catalog No. NP233194A30
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CD19
C19/366
Immunohistochemistry, Immunohistochemistry-Paraffin
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD19
A recominant protein fragment specific to CD19 was used as the immunogen for the C19/366 antibody.
0.1 mL
Primary
930