Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Mouse anti-Human, Alexa Fluor™ 350, Clone: HD37, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252705AF350

Catalog No. NP2270A30


Add to cart

Description

Description

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunofluorescence.
Specifications

Specifications

CD19
HD37
Flow Cytometry, Immunohistochemistry, Immunofluorescence
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Recognises human CD19.
Flow Cytometry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CD19
Cells from a hairy cell leukemia patient.
0.1 mL
Primary
930
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit