Learn More
CD19 Mouse anti-Human, Alexa Fluor™ 532, Clone: FMC63, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP252716AF532
Catalog No. NP22716A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunofluorescence.
Specifications
Specifications
CD19
FMC63
Flow Cytometry, Immunohistochemistry, Immunofluorescence
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Binds human CD19.
Flow Cytometry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD19
Human prolymphocytic leukaemia cell line JVM3.
0.1 mL
Primary
930
Human