Learn More
CD19 Mouse anti-Human, Alexa Fluor™ 750, Clone: Bu12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10064360AF750
Catalog No. N1064360A70
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD19
Bu12
Flow Cytometry
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
NB100-64360 recognizes human CD19, a 95kD cell surface glycoprotein, which is expressed on cells of the B cell lineage and follicular dendritic cells but absent on plasma cells. CD19 is an important signal transduction molecule which is involved in the regulation of B lymphocyte development, activation and differentiation.
Flow Cytometry
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CD19
Human EB-4 Burkitt lymphoma cell line.
0.1 mL
Primary
930
Human