Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, BB700, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
$140.00 - $384.00
Specifications
Antigen | CD19 |
---|---|
Clone | SJ25C1 (also known as SJ25-C1) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry.Specifications
CD19 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Mouse | |
Human | |
Primary |
SJ25C1 (also known as SJ25-C1) | |
Monoclonal | |
Purified | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title