Learn More
CD19 Mouse anti-Human, Brilliant Ultraviolet 496, Clone: SJ25C1 (also known as SJ25-C1), BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 612938
 View more versions of this product
Catalog No. BDB612938
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD19
SJ25C1 (also known as SJ25-C1)
Aqueous buffered solution containing ≤0.09% sodium azide.
B4; B-lymphocyte antigen CD19; Leu-12
Human NALM1 + NALM16 cells
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Brilliant Ultraviolet 496
Affinity Purified
Mouse
BALB/c IgG1, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal