Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD19 Mouse anti-Human, Monkey, DyLight 755, Clone: CVID3/155, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261908IR
Description
CD19 Monoclonal antibody specifically detects CD19 in Human, Monkey samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunofluorescence.Specifications
CD19 | |
CVID3/155 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Conjugated, Purified | |
CD19 | |
Recombinant human CD19 protein | |
Protein A or G purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
Primary | |
930 | |
Human, Monkey |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12 | |
Mouse | |
IgG1 Kappa | |
0.1 ml | |
Store at 4°C in the dark. | |
Monoclonal | |
Recognizes a protein of 95kDa, identified as CD19. CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title