Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Mouse anti-Human, Monkey, Primate, PE, Clone: CVID3/429, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261909PE

Catalog No. NBP261909PE


Add to cart

Description

Description

CD19 Monoclonal antibody specifically detects CD19 in Human, Monkey, Primate samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD19
CVID3/429
Flow Cytometry
Conjugated, Purified
CD19
Recombinant human CD19 protein
Protein A or G purified
Adaptive Immunity, B Cell Development and Differentiation Markers, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary
930
Human, Monkey, Primate
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1 Kappa
0.1 ml
Store at 4°C in the dark.
Monoclonal
CD19 is a transmembrane glycoprotein that contains two extracellular immunoglobulin-like domains. CD19 is present in both benign and malignant B-cells and is considered to be the most reliable surface marker of this lineage over a wide range of maturational stages. In normal lymphoid tissue, CD19 is observed in germinal centers, in mantle zone cells, and in scattered cells of the inter-follicular areas. Anti-CD19 exhibits an overall immunoreactivity pattern similar to those of the antibodies against CD20 and CD22. However, in contrast to CD20, expression of CD19 is continuous throughout B-cell development and through terminal differentiation of B-cells into plasma cells. Anti-CD19 positivity is seen in the vast majority of B-cell neoplasms commonly at a lower intensity than normal B-cell counterparts. Plasma cell neoplasms are nearly always negative, as are T-cell neoplasms
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit