Learn More
CD19 Mouse anti-Human, Mouse, Alexa Fluor™ 532, Clone: CB19, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225196AF532
Catalog No. NP22196A32
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human,Mouse samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
CD19
CB19
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CD19
Intact normal human B cell cells were used as immunogen to generate the CB19 clone.
0.1 mL
Primary
930