Learn More
CD19 Mouse anti-Human, PE, Clone: COC19, Abnova™
Mouse monoclonal antibody raised against human CD19.
Manufacturer:  Abnova Corporation MAB12398
Catalog No. 89-933-753
Add to cart
Description
Description
Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. This gene encodes a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. [provided by RefSeq]
Specifications
Specifications
CD19
COC19
Mouse monoclonal antibody raised against human CD19.
In PBS, pH 7.4 (0.09% sodium azide, 0.2% BSA)
B4/MGC12802
Mouse
IgG1
50 Reactions
Store at 4°C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunofluorescence
PE
Flow cytometry Immunofluorescence The optimal working dilution should be determined by the end user.
Liquid
CD19
CD19
Protein A/G affinity chromatography
RUO
Primary
930