Learn More
CD19 Mouse anti-Human, PE-Cyanine7, Clone: SJ25-C1, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD1912
Catalog No. 50-113-7504
Add to cart
Description
Description
The R-phycoerythrin (PE)-Texas Red™ conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD19 is a member of the immunoglobulin superfamily and has two Ig like domains. The CD19 molecule is expressed on 100% of the peripheral B cells as defined by expression of kappa or lambda light chains. CD19 appears to be expressed on myeloid leukemia cells, particularly those of monocytic lineage. Leukemia phenotype studies have demonstrated that the earliest and broadest B cell restricted antigen is the CD19 antigen. The receptor for CD19 is an important functional regulator of normal and maligt B cell proliferation, and is expressed in all B cell precursor leukemias. Lymphocytes proliferate and differentiate in response to various concentrations of different antigens. The ability of the B cell to respond in a specific, yet sensitive manner to the various antigens is achieved with the use of low-affinity antigen receptors. CD19 is a cell surface molecule which assembles with the antigen receptor of B lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. Besides being a signal-amplifying coreceptor for the B cell receptor (BCR), CD19 can also signal independently of BCR co-ligation and is a central regulatory component upon which multiple signaling pathways converge. Mutation of the CD19 gene results in hypogammaglobulinemia, whereas CD19 overexpression causes B cell hyperactivity.
Specifications
Specifications
CD19
Monoclonal
PE-Cyanine7
CD19
Liquid
CD19
Nalm 1 Human cell line
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
SJ25-C1
PBS with 4mg/mL BSA and 0.1% sodium azide
P15391
AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19
Mouse
IgG1, kappa
0.5 mL
4° C, store in dark
Primary
930