Learn More
CD19 Rat anti-Human, Mouse, Alexa Fluor™ 750, Clone: 1D3, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP224965AF750
Catalog No. NP22496A70
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human,Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD19
1D3
Flow Cytometry, Flow (Cell Surface)
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CD19
Mouse CD19 Transfected Cell Line
0.1 mL
Primary
930