Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD193 Rat anti-Mouse, BB700, Clone: 83103, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 742079

Catalog No. BDB742079


Add to cart

Description

Description

The 83103 monoclonal antibody specifically binds to CD193, which is also known as, C-C chemokine receptor type 3 (CCR3). CD193 is a chemokine receptor for CCL11/eotaxin, CCL24/eotaxin-2, CCL26/eotaxin-3, CCL5/RANTES, CCL7/MCP-3, CCL8/MCP-2, CCL14/MCP-4, CCL15/HCC-2 and CCL3/MIP-1a. CD193 is reportedly expressed on eosinophils, mast cells and Th2 cells. It plays an important role in eosinophil recruitment to the skin and the lung and in the development of airway hyperresponsiveness.

The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5.  Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.
Specifications

Specifications

CD193
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
CCR3, CCR-3, chemokine (C-C) receptor 3, CC-CKR3, C-C CKR-3, CKR3, CC-CKR-3
Mouse CCR3 transfected Cells Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BB700 under optimal conditions that minimize unconjugated dye and antibody.
RUO
Primary
Murine
83103
BB700
Affinity Purified
Rat
Rat IgG2a
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit