Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD196 (CCR6) Rat anti-Mouse, BV480, Clone: 140706, BD OptiBuild
Rat Monoclonal Antibody
Manufacturer: BD Biosciences 747830
Description
CD196 (CCR6) Monoclonal antibody specifically detects CD196 (CCR6) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD196 (CCR6) | |
140706 | |
Brilliant Violet 480 | |
Rat | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG2a | |
RUO | |
Primary | |
Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title