Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD197, CCR7 Rat anti-Human, BB515, Clone: 3D12, BD Horizon™

Rat Monoclonal Antibody

$118.00 - $242.00

Specifications

Antigen CD197, CCR7
Isotype Rat IgG2a, κ
Immunogen Human CCR7 protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Conjugate BB515
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB565870
View Documents
BD Biosciences
565870
25 Tests
Each for $118.00
Add to cart
 
BDB565869
View Documents
BD Biosciences
565869
100 Tests
Each for $242.00
Add to cart
 
Description

Description

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.

CD197, CCR7 Monoclonal antibody specifically detects CD197, CCR7 in Human samples. It is validated for Flow cytometry.
Specifications

Specifications

CD197, CCR7
Human CCR7 protein
BB515
Cell Biology
Rat
Purified
Monoclonal
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Rat IgG2a, κ
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Flow Cytometry
RUO
3D12
CCR7, BLR-2, EBI-1, CMKBR7
Vol. Per test: 5μL
SDS
Product Certifications

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit