Learn More
CD199 (CCR9) Mouse anti-Mouse, BB515, Clone: CW-1.2, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565577
Catalog No. BDB565577
Add to cart
Description
Description
CD199 (CCR9) Monoclonal antibody specifically detects CD199 (CCR9) in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD199 (CCR9)
CW-1.2
BB515
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Conjugated, Purified
IgG2a, κ
RUO
Primary
CD199 (CCR9)