Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD1a Ab-5, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD1a Ab-5, Mouse Monoclonal Antibody.

$207.50 - $1135.00

Specifications

Antigen CD1a Ab-5
Clone O10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 8
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1856P
View Documents
Thermo Scientific™
MS1856P
Purified Ab with BSA and Azide 1mL
Each for $1,085.00
Add to cart
 
MS1856P0
View Documents
Thermo Scientific™
MS1856P0
Purified Ab with BSA and Azide 100μL
Each for $207.50
Add to cart
 
MS1856P1
View Documents
Thermo Scientific™
MS1856P1
Purified Ab with BSA and Azide 500μL
Each for $571.50
Add to cart
 
MS1856P1ABX
View Documents
Thermo Scientific™
MS1856P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $617.50
Add to cart
 
MS1856PABX
View Documents
Thermo Scientific™
MS1856PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $1,135.00
Add to cart
 
MS1856R7
View Documents
Thermo Scientific™
MS1856R7
Ready-to-use for IHC (UVLP) 7mL
Each for $490.50
Add to cart
 
MS1856RQ
View Documents
Thermo Scientific™
MS1856RQ
Ready-to-use for IHC (Quanto) 12mL
Each for $531.50
Add to cart
 
MS1856R1
View Documents
Thermo Scientific™
MS1856R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Obtain accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD1a Ab-5, Mouse Monoclonal Antibody.

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: O10

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Molecular Weight: 49kDa

Positive Control: Tonsil

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD1a Ab-5
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
O10
Monoclonal
Murine
Hematopoietic
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit