Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1A Monoclonal Antibody (A3F9), Huabio
Mouse Monoclonal Antibody
Supplier: HUABIO RESEARCH INC EM190189


Description
CD1A Monoclonal antibody specifically detects CD1A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
CD1A | |
Monoclonal | |
2 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P06126 | |
Mouse | |
Protein G | |
RUO | |
909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
A3F9 | |
Unconjugated | |
CD1A | |
CD1; CD1 antigen; CD1.1; CD1A; CD1A antigen, a polypeptide; CD1a molecule; cd1a1; CD1A1 antigen; cluster of differentiation 1 A; cortical thymocyte antigen CD1A; differentiation antigen CD1-alpha-3; epidermal dendritic cell marker CD1a; FCB6; HTA1; hTa1 thymocyte antigen; R4; T6; T-cell surface antigen T6/Leu-6; T-cell surface glycoprotein CD1a | |
Recombinant protein within human CD1A aa 50-250. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction