Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1A Mouse anti-Human, Clone: CBT6, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB15052

Catalog No. 89-035-072


Add to cart

Description

Description

Mouse monoclonal antibody raised against native human CD1A.

  • High specificity
  • High reactivity
Specifications

Specifications

CD1A
CBT6
Mouse monoclonal antibody raised against native human CD1A.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
CD1A
Native purified human CD1A.
Protein A/G purification
RUO
Primary
909
Flow Cytometry, Immunofluorescence
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user.
P06126
CD1|FCB6|HTA1|R4|T6
Mouse
IgG2b, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)