Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1a Rabbit anti-Human, Allophycocyanin/Cy7, Clone: C1A/1506R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254547APCCY7

Catalog No. N254547APC7


Add to cart

Description

Description

CD1a Monoclonal antibody specifically detects CD1a in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD1a
C1A/1506R
Flow Cytometry
Conjugated, Purified
CD1A
Recombinant human full-length CD1a protein
Protein A purified
Dendritic Cell Markers, Immunology
Primary
909.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a
Rabbit
IgG Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit