Learn More
CD1b Mouse anti-Human, Janelia Fluor 549, Clone: SN13, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143459JF549
Catalog No. NBP43459549
Add to cart
Description
Description
CD1b Monoclonal antibody specifically detects CD1b in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD1b
SN13
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD1B
The immunogen for this antibody was CD1b.
Protein A or G purified
Immunology
Primary
910
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD1, CD1A, CD1b antigen, CD1B antigen, b polypeptide, CD1b molecule, cortical thymocyte antigen CD1B, differentiation antigen CD1-alpha-3, MGC125990, MGC125991, R1, T-cell surface glycoprotein CD1b
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human