Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1c/BDCA-1 Mouse anti-Human, DyLight™ 350, Clone: OTI2F4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270345UV
Description
CD1c/BDCA-1 Monoclonal antibody specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CD1c/BDCA-1 | |
OTI2F4 | |
Western Blot, Immunohistochemistry | |
50mM Sodium Borate with 0.05% Sodium Azide | |
911.0 | |
Mouse | |
IgG1 | |
Primary | |
0.1 mL | |
Human |
Western Blot,Immunohistochemistry | |
DyLight 350 | |
Affinity Purified,Conjugated | |
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c | |
CD1C | |
Full length recombinant protein of human CD1c/BDCA-1 (NP_001756) produced in HEK293T cells. | |
Monoclonal | |
Immunogen affinity purified | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title