Learn More
CD1c/BDCA-1 Antibody (OTI2F4), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270345C
Catalog No. NBP270345C
Add to Cart
Description
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
CD1c/BDCA-1
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
CD1C
Full length recombinant protein of human CD1c/BDCA-1 (NP_001756) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI2F4
Western Blot, Immunohistochemistry
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
Mouse
Affinity Purified
Cancer, Immunology
911.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions