Learn More
CD1c/BDCA-1 Mouse anti-Human, Alexa Fluor™ 700, Clone: L161, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262220AF700
Catalog No. NB002728
Add to cart
Description
Description
CD1c/BDCA-1 Monoclonal specifically detects CD1c/BDCA-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
CD1c/BDCA-1
L161
Flow Cytometry, Immunohistochemistry
Purified, Conjugated
CD1C
human thymocytes
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody MEM-97 reacts with CD20 (Bp35), a 33-37 kDa non-glycosylated membrane receptor with four transmembrane domains, expressed on B lymphocytes (it is lost on plasma cells), follicular dendritic cells, and at low levels on peripheral blood T lymphocytes.
Flow Cytometry, Immunohistochemistry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
BDCA1, c polypeptide, CD1, CD1A, CD1c antigen, CD1c molecule, cortical thymocyte antigen CD1C, differentiation antigen CD1-alpha-3, R7, T-cell surface glycoprotein CD1c
Mouse
IgG1 Kappa
0.1 mL
Primary
911
Human