Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD1C Mouse anti-Human, Clone: UMAB46, liquid, UltraMAB™

Mouse Monoclonal Antibody
Supplier: OriGene UM570042

Description
This gene encodes a member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by this gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail, and requires vesicular acidification to bind lipid antigens.Specifications
CD1C | |
Monoclonal | |
0.5-1.0 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P29017 | |
CD1C | |
Full length human recombit protein of human CD1C produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin) | |
UMAB46 | |
Unconjugated | |
CD1C | |
BDCA1, CD1, CD1A, R7 | |
Mouse | |
Affinity Chromatography | |
RUO | |
911 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title