Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d Mouse anti-Mouse, Rat, APC, Clone: WTH-2, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 17001380

Catalog No. 50-112-3022


Add to cart

Description

Description

Description: The monoclonal antibody WTH-2 reacts with mouse and rat CD1d. CD1d is a non-polymorphic MHC Class I-like molecule composed of alpha-1 and alpha-2 domains. CD1d is constitutively expressed on hematopoietic cells, with highest levels on dendritic cells, macrophages, and marginal zone B-cells. It is also expressed on non-lymphatic organs such as the liver, heart, small intestine, and pancreas. CD1d associates with beta2-microglobulin to present antigens containing a lipid or other hydrophobic moieties to invariant NKT cells, which leads to rapid Th1 or Th2 cytokine secretion. The WTH-2 antibody has been reported to have antagonist activity. Applications Reported: This WTH-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This WTH-2 antibody has been tested by flow cytometric analysis of mouse or rat splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD1d is a 49 kDa heavy chain associated with Beta 2 microglobulin on cortical thymocytes. Beta 2 microglobulin independent expression of CD1d has also been demonstrated on human intestinal epithelial cells. CD1d is the sole group 2 member of the CD1 family of major histocompatibility (MHC) like glycoproteins. The CD1d gene encodes a divergent member of the CD1 family of transmembrane glycoproteins, which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin. The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells. The human genome contains five CD1 family genes organized in a cluster on chromosome 1. The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands. The protein encoded by the CD1d gene localizes to late endosomes and lysosomes via a tyrosine-based motif in the cytoplasmic tail. Diseases associated with CD1D include Mycobacterium Malmoense and Autoimmune Disease.
Specifications

Specifications

CD1d
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P11609, Q63493
AI747460, CD1.1, Cd1a, Cd1d, Ly-38, T-cell surface glycoprotein CD1d1, antigen-presenting glycoprotein CD1d1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse, Rat
Flow Cytometry
WTH-2
APC
Cd1d1
Liquid
Cd1d1
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12479, 25109
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.