Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD1d, PerCP-eFluor™ 710, clone: 1B1, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

$138.00 - $321.00

Specifications

Antigen CD1d
Clone 1B1
Target Species Murine
Conjugate PerCP-eFluor 710
Isotype IgG2b, kappa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-113-0729 Non-distribution product as customer accommodation. View Documents Invitrogen
46001180
25μg Each for $138.00
Add to cart
 
50-113-0730 Non-distribution product as customer accommodation. View Documents Invitrogen
46001182
100μg Each for $321.00
Add to cart
 
Description

Description

CD1d is a 49kD heavy chain associated with Beta 2 microglobulin on cortical thymocytes. Beta 2 microglobulin independent expression of CD1d has also been demonstrated on human intestinal epithelial cells. CD1D is the sole group 2 member of the CD1 family of major histocompatibility (MHC) like glycoproteins.

Description: The 1B1 monoclonal antibody reacts with mouse CD1d, a 48kDa glycoprotein with structural homology to MHC class I molecules. While similar to MHC Class I, CD1d associates with beta2m, functionally CD1d is similar to MHC Class II. 1B1 detects CD1d at varying levels on mouse leukocytes. 1B1 detects beta2m associated CD1d. Applications Reported: This 1B1 antibody has been reported for use in flow cytometric analysis. Applications Tested: %This 1B1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.%.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2µm post-manufacturing filtered.
Specifications

Specifications

CD1d
Murine
IgG2b, kappa
Monoclonal
Liquid
Antibody
Store at 2-8°C. Do not freeze. Light-sensitive material.
1B1
PerCP-eFluor 710
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit