Learn More
ImmudexSupplier Diversity Partner Mouse CD1d 150 tests DFS Item
Manufacturer:  ImmudexSupplier Diversity Partner YD8002
CD1d Dextramer can identify NKT cells in fluid cell samples including blood, cultured cell lines, CSF, lymph, synovial fluid using flow cytometry. CD1d Dextramer carry multiple CD1d molecules which are non-classical MHC molecules with hydrophobic binding pockets wherein glycolipids and phospholipids are presented. The CD1d-lipid complexes binds to T-cell receptors on a subset of NKT cells (CD1d-restricted NKT cells).
Catalog No. NC1472953